Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130914.1 | NP_001124386.1 | 5, 6 | 916 | ||
NM_006806.4 | NP_006797.3 | 4, 5 | 784 | |||
XM_011529441.1 | XP_011527743.1 | 6, 7 | 1024 | |||
XM_017028265.1 | XP_016883754.1 | 5, 6 | 922 | |||
XM_017028266.1 | XP_016883755.1 | 5, 6 | 879 | |||
GenBank mRNA | AK289408.1 | - | 4, 5 | 732 | ||
BC011957.1 | - | 5, 6 | 810 | 3859781 | ||
BT007276.1 | - | 4, 5 | 656 | |||
CR457061.1 | - | 3, 4 | 524 | |||
D64110.1 | - | 4, 5 | 618 | |||
DR155504.1 | - | 4, 5 | 700 |