Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006808.2 | NP_006799.1 | 3 | 267 | ||
GenBank mRNA | AJ293399.1 | - | 2 | 132 | ||
AK223384.1 | - | 3 | 278 | |||
BC001734.1 | - | 3 | 267 | 3502133 | ||
CR456883.1 | - | 2 | 191 | |||
CR542159.1 | - | 2 | 191 | |||
L25085.1 | - | 3 | 254 |