Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017956.2 | NP_001017956.1 | 8 | 1124 | ||
NM_001017957.2 | NP_001017957.1 | 8 | 1124 | |||
NM_001017958.2 | NP_001017958.1 | 8 | 1124 | |||
NM_001261420.1 | NP_001248349.1 | 8 | 1127 | |||
NM_001261421.1 | NP_001248350.1 | 7 | 1028 | |||
NM_001261422.1 | NP_001248351.1 | 7 | 968 | |||
NM_001261423.1 | NP_001248352.1 | 7 | 947 | |||
NM_006812.3 | NP_006803.1 | 8 | 1124 | |||
XM_005268581.1 | XP_005268638.1 | 8 | 1059 | |||
XM_006719200.1 | XP_006719263.1 | 8 | 1059 | |||
XM_006719201.1 | XP_006719264.1 | 8 | 1059 | |||
GenBank mRNA | AB002806.1 | - | 8 | 958 | ||
AK129781.1 | - | 8 | 939 | |||
AK130733.1 | - | 8 | 937 | |||
AK293435.1 | - | 7 | 759 | |||
AK296770.1 | - | 8 | 934 | |||
AK298532.1 | - | 7 | 838 | |||
AK303291.1 | - | 7 | 1311 | |||
AK303858.1 | - | 7 | 778 | |||
AL137691.1 | - | 4 | 1219 | |||
BC000532.2 | - | 8 | 929 | 2964645 | ||
BC006506.2 | - | 1 | 37 | |||
BC007254.1 | - | 8 | 929 | 3027883 | ||
BC023513.2 | - | 8 | 919 | 4123782 | ||
CR456791.1 | - | 8 | 917 | |||
EF036505.1 | - | 2 | 52 | |||
U41635.1 | - | 8 | 1002 |