Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006816.2 | NP_006807.1 | 1 | 370 | ||
GenBank mRNA | AK222609.1 | - | 1 | 137 | ||
AK292099.1 | - | 1 | 147 | |||
AK301607.1 | - | 2 | 54 | |||
AK308027.1 | - | 1 | 281 | |||
BC017263.2 | - | 1 | 150 | 4135210 | ||
DC404202.1 | - | 1 | 370 | |||
U10362.1 | - | 1 | 133 |