Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI44L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006820.3 | NP_006811.2 | 2, 3 | 687 | ||
XM_005270391.3 | XP_005270448.1 | 1, 2 | 194 | |||
XM_005270392.4 | XP_005270449.1 | 1, 2 | 170 | |||
XM_006710304.3 | XP_006710367.1 | 2, 3 | 197 | |||
XM_011540539.2 | XP_011538841.1 | 2, 3 | 525 | |||
XM_017000120.1 | XP_016855609.1 | 2, 3 | 195 | |||
GenBank mRNA | AB000115.1 | - | 2, 3 | 611 | ||
AK223087.1 | - | 2, 3 | 621 | |||
AK225442.1 | - | 2, 3 | 588 | |||
AK293385.1 | - | 2, 3 | 286 | |||
AK316232.1 | - | 1, 2 | 136 | |||
AL832618.2 | - | 1 | 3290 | |||
BC015932.2 | - | 2, 3 | 593 | 3909331 | ||
DC390321.1 | - | 1, 2 | 199 |