Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI44L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006820.3 | NP_006811.2 | 8 | 1383 | ||
XM_005270391.3 | XP_005270448.1 | 7 | 890 | |||
XM_005270392.4 | XP_005270449.1 | 6 | 670 | |||
XM_005270393.4 | XP_005270450.1 | 5 | 592 | |||
XM_006710304.3 | XP_006710367.1 | 7 | 697 | |||
XM_011540539.2 | XP_011538841.1 | 8 | 1221 | |||
XM_017000120.1 | XP_016855609.1 | 8 | 891 | |||
GenBank mRNA | AB000115.1 | - | 8 | 1307 | ||
AK223087.1 | - | 8 | 1317 | |||
AK225442.1 | - | 8 | 1284 | |||
AK293385.1 | - | 8 | 982 | |||
AK303187.1 | - | 5 | 584 | |||
AK312272.1 | - | 8 | 1290 | |||
AK316232.1 | - | 7 | 832 | |||
AL832618.2 | - | 6 | 3986 | |||
BC015932.2 | - | 8 | 1289 | 3909331 |