Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006826.3 | NP_006817.1 | 2, 3 | 499 | ||
GenBank mRNA | AF070556.1 | - | 1, 2 | 88 | ||
AK297624.1 | - | 3, 4 | 332 | |||
AK302208.1 | - | 2, 3 | 205 | |||
AK309462.1 | - | 3, 4 | 328 | |||
AK316158.1 | - | 3, 4 | 327 | |||
BC001197.1 | - | 436 | ||||
BC050601.1 | - | 1, 2 | 312 | 6501365 | ||
BC056867.1 | - | 2, 3 | 434 | 6164592 | ||
BC093019.1 | - | 2, 3 | 441 | 30520472 | ||
BT020014.1 | - | 1, 2 | 302 | |||
X56468.1 | - | 2, 3 | 427 | |||
X57347.1 | - | 2, 3 | 402 |