Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006826.3 | NP_006817.1 | 3 | 604 | ||
GenBank mRNA | AF070556.1 | - | 2 | 193 | ||
AK297624.1 | - | 4 | 437 | |||
AK302208.1 | - | 3 | 310 | |||
AK309462.1 | - | 4 | 433 | |||
AK316158.1 | - | 4 | 432 | |||
BC001197.1 | - | 541 | ||||
BC050601.1 | - | 2 | 417 | 6501365 | ||
BC056867.1 | - | 3 | 539 | 6164592 | ||
BC093019.1 | - | 3 | 546 | 30520472 | ||
X56468.1 | - | 3 | 532 | |||
X57347.1 | - | 3 | 507 |