Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006836.1 | NP_006827.1 | 14 | 1259 | ||
GenBank mRNA | AK297271.1 | - | 14 | 1249 | ||
BC153881.1 | - | 14 | 1259 | 8860118 | ||
BQ929459.1 | - | 5 | 359 | |||
D86973.2 | - | 14 | 1259 |