Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006837.2 | NP_006828.2 | 3 | 785 | ||
GenBank mRNA | AB209134.1 | - | 2 | 2495 | ||
AK056967.1 | - | 5 | 561 | |||
AK128519.1 | - | 2 | 2777 | |||
AK310434.1 | - | 3 | 587 | |||
AK311071.1 | - | 3 | 589 | |||
BC001187.1 | - | 3 | 589 | 3354130 | ||
BC001859.2 | - | 3 | 567 | 3530890 | ||
BC007272.1 | - | 3 | 563 | 3161025 | ||
BX648542.1 | - | 3 | 3122 | |||
CR541678.1 | - | 3 | 454 | |||
U65928.1 | - | 3 | 581 | |||
U70734.1 | - | 3 | 574 |