Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317182.1 | NP_001304111.1 | 5 | 631 | ||
NM_001317183.1 | NP_001304112.1 | 4 | 631 | |||
NM_006838.3 | NP_006829.1 | 5 | 700 | |||
NR_133673.1 | 5 | 700 | ||||
XM_005268583.3 | XP_005268640.1 | 5 | 619 | |||
GenBank mRNA | AK055692.1 | - | 422 | |||
AK091730.1 | - | 4 | 507 | |||
AK125296.1 | - | 5 | 1226 | |||
AK300836.1 | - | 5 | 596 | |||
AK315559.1 | - | 5 | 637 | |||
BC013782.1 | - | 5 | 578 | 4340717 | ||
U13261.1 | - | 5 | 600 |