Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100169.1 | NP_001093639.1 | 15 | 2217 | ||
NM_001100170.1 | NP_001093640.1 | 15 | 2187 | |||
NM_006839.2 | NP_006830.2 | 15 | 2220 | |||
XM_005264110.3 | XP_005264167.1 | 15 | 1856 | |||
XM_005264113.1 | XP_005264170.1 | 14 | 1792 | |||
XM_005264114.1 | XP_005264171.1 | 14 | 1763 | |||
XM_011532497.2 | XP_011530799.1 | 15 | 1880 | |||
XM_017003194.1 | XP_016858683.1 | 15 | 1888 | |||
XM_017003195.1 | XP_016858684.1 | 15 | 1912 | |||
GenBank mRNA | AK225780.1 | - | 15 | 1854 | ||
AK296676.1 | - | 15 | 1826 | |||
AK299014.1 | - | 14 | 1742 | |||
AK299596.1 | - | 13 | 1597 | |||
AK300017.1 | - | 13 | 1563 | |||
AK312251.1 | - | 15 | 1870 | |||
AY232292.1 | - | 15 | 1850 | |||
AY921637.1 | - | 15 | 1868 | |||
BC002412.2 | - | 15 | 1861 | 2964676 | ||
BC027458.1 | - | 4 | 573 | |||
BX537369.1 | - | 15 | 1891 | |||
CR456793.1 | - | 15 | 1832 | |||
CR749590.1 | - | 10 | 1264 | |||
D21092.1 | - | 13 | 1722 | |||
D21094.1 | - | 15 | 1917 | |||
L42572.1 | - | 15 | 1924 |