Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100169.1 | NP_001093639.1 | 10 | 1490 | ||
NM_001100170.1 | NP_001093640.1 | 10 | 1460 | |||
NM_006839.2 | NP_006830.2 | 10 | 1493 | |||
XM_005264110.3 | XP_005264167.1 | 10 | 1129 | |||
XM_005264113.1 | XP_005264170.1 | 9 | 1065 | |||
XM_005264114.1 | XP_005264171.1 | 9 | 1036 | |||
XM_011532497.2 | XP_011530799.1 | 10 | 1153 | |||
XM_017003194.1 | XP_016858683.1 | 10 | 1161 | |||
XM_017003195.1 | XP_016858684.1 | 10 | 1185 | |||
GenBank mRNA | AK225780.1 | - | 10 | 1127 | ||
AK296676.1 | - | 10 | 1099 | |||
AK299014.1 | - | 9 | 1015 | |||
AK299596.1 | - | 8 | 870 | |||
AK300017.1 | - | 8 | 836 | |||
AK312251.1 | - | 10 | 1143 | |||
AY232292.1 | - | 10 | 1123 | |||
AY921637.1 | - | 10 | 1141 | |||
BC002412.2 | - | 10 | 1134 | 2964676 | ||
BX537369.1 | - | 10 | 1163 | |||
CR456793.1 | - | 10 | 1105 | |||
CR749590.1 | - | 5 | 537 | |||
D21092.1 | - | 8 | 995 | |||
D21094.1 | - | 10 | 1190 | |||
L42572.1 | - | 10 | 1197 |