Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3AR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326475.1 | NP_001313404.1 | 2 | 994 | ||
NM_001326477.1 | NP_001313406.1 | 2 | 1054 | |||
NM_004054.3 | NP_004045.1 | 2 | 997 | |||
GenBank mRNA | AK223388.1 | - | 2 | 907 | ||
AK290242.1 | - | 2 | 885 | |||
AK312458.1 | - | 2 | 885 | |||
BC020742.1 | - | 2 | 885 | 4690283 | ||
U28488.1 | - | 2 | 874 | |||
U62027.1 | - | 2 | 945 | |||
Z73157.1 | - | 1 | 793 |
Set Membership: |
G-protein coupled receptor |