Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC27A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321196.1 | NP_001308125.1 | 4 | 990 | ||
NM_012254.2 | NP_036386.1 | 4 | 1242 | |||
XM_011526364.1 | XP_011524666.1 | 4 | 1171 | |||
XM_017026214.1 | XP_016881703.1 | 4 | 1171 | |||
GenBank mRNA | AF064255.1 | - | 4 | 1202 | ||
AK123036.1 | - | 4 | 1242 | |||
AK298446.1 | - | 4 | 917 |