Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC27A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005094.3 | NP_005085.2 | 2 | 437 | ||
XM_017014222.1 | XP_016869711.1 | 3 | 310 | |||
GenBank mRNA | AF055899.1 | - | 1 | 152 | ||
AK290222.1 | - | 2 | 437 | |||
AK314681.1 | - | 2 | 277 | |||
BC065003.1 | - | 2 | 436 | 6023438 |