Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297655.1 | NP_001284584.1 | 9 | 902 | ||
NM_001297656.1 | NP_001284585.1 | 9 | 1143 | |||
NM_001297657.1 | NP_001284586.1 | 9 | 830 | |||
NM_006845.3 | NP_006836.2 | 10 | 1025 | |||
XM_011540540.2 | XP_011538842.1 | 10 | 1272 | |||
XM_011540541.2 | XP_011538843.1 | 9 | 1213 | |||
GenBank mRNA | AB264115.1 | - | 10 | 1025 | ||
AK290688.1 | - | 10 | 951 | |||
AK300756.1 | - | 9 | 859 | |||
AK302267.1 | - | 9 | 798 | |||
AK312606.1 | - | 10 | 981 | |||
AY026505.1 | - | 9 | 1096 | |||
BC008764.2 | - | 10 | 964 | 3347571 | ||
BC014924.1 | - | 10 | 962 | 3909438 | ||
BC032038.1 | - | 2 | 140 | |||
BT006759.1 | - | 10 | 910 | |||
CR450302.1 | - | 10 | 910 | |||
U63743.1 | - | 10 | 964 |