Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297655.1 | NP_001284584.1 | 1, 2 | 180 | ||
NM_006845.3 | NP_006836.2 | 1, 2 | 180 | |||
XM_011540540.2 | XP_011538842.1 | 1, 2 | 294 | |||
GenBank mRNA | AB264115.1 | - | 1, 2 | 180 | ||
AK290688.1 | - | 1, 2 | 106 | |||
AK300756.1 | - | 1, 2 | 137 | |||
AU130795.1 | - | 1, 2 | 180 | |||
BC008764.2 | - | 1, 2 | 119 | 3347571 | ||
BC014924.1 | - | 1, 2 | 117 | 3909438 | ||
BT006759.1 | - | 1, 2 | 65 | |||
CR450302.1 | - | 1, 2 | 65 | |||
U63743.1 | - | 1, 2 | 119 |