Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278426.3 | NP_001265355.2 | 12 | 1715 | ||
NM_001278427.3 | NP_001265356.2 | 12 | 1712 | |||
NM_001278428.3 | NP_001265357.2 | 12 | 1715 | |||
NM_001278429.3 | NP_001265358.2 | 11 | 1682 | |||
XM_017026215.1 | XP_016881704.1 | 13 | 1742 | |||
XM_017026216.1 | XP_016881705.1 | 12 | 1715 | |||
XM_017026217.1 | XP_016881706.1 | 12 | 1709 | |||
GenBank mRNA | AF025532.1 | - | 12 | 1529 | ||
AF283988.1 | - | 12 | 1536 | |||
AK292082.1 | - | 12 | 1579 | |||
BC026309.1 | - | 12 | 1534 | 4795550 | ||
U91925.1 | - | 12 | 1545 |