Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006851.2 | NP_006842.2 | 3 | 616 | ||
XM_011537775.2 | XP_011536077.1 | 4 | 666 | |||
XM_017018721.1 | XP_016874210.1 | 3 | 496 | |||
GenBank mRNA | AF400440.1 | - | 3 | 496 | ||
AK313449.1 | - | 3 | 482 | |||
BC012510.1 | - | 3 | 532 | 4453841 | ||
EF667354.1 | - | 4 | 539 | |||
U16307.3 | - | 3 | 552 | |||
X91911.1 | - | 3 | 596 |