Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136032.2 | NP_001129504.1 | 6 | 1009 | ||
NM_001167605.1 | NP_001161077.1 | 6 | 1096 | |||
NM_006853.2 | NP_006844.1 | 6 | 1021 | |||
NM_144947.1 | NP_659196.1 | 6 | 1103 | |||
XM_005258439.3 | XP_005258496.1 | 4 | 742 | |||
XM_011526369.1 | XP_011524671.1 | 6 | 1247 | |||
XM_011526370.2 | XP_011524672.1 | 6 | 1121 | |||
XM_011526371.2 | XP_011524673.1 | 7 | 1241 | |||
XM_011526372.2 | XP_011524674.1 | 6 | 1154 | |||
XM_011526373.1 | XP_011524675.1 | 6 | 1024 | |||
GenBank mRNA | AB012917.1 | - | 6 | 1016 | ||
AB013730.1 | - | 6 | 1015 | |||
AB041036.1 | - | 6 | 1103 | |||
AB259014.1 | - | 6 | 1007 | |||
AY359014.1 | - | 6 | 1000 | |||
BC022068.1 | - | 6 | 1027 | 4824387 |