Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMSB15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021992.2 | NP_068832.1 | 3 | 446 | ||
GenBank mRNA | AF086495.1 | - | 3 | 389 | ||
BC000183.1 | - | 3 | 394 | 3350388 | ||
BG829126.1 | - | 3 | 396 | |||
BU599402.1 | - | 3 | 459 | |||
D82345.1 | - | 3 | 420 |