Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006857.2 | NP_006848.1 | 6 | 467 | ||
NR_037862.1 | 6 | 467 | ||||
GenBank mRNA | AK291128.1 | - | 6 | 448 | ||
AK312041.1 | - | 6 | 432 | |||
BC017890.1 | - | 6 | 452 | 4699198 | ||
BU171817.1 | - | 6 | 458 | |||
X76302.1 | - | 6 | 451 |