Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006858.3 | NP_006849.1 | 4 | 904 | ||
NR_104015.1 | 4 | 743 | ||||
XM_006722631.3 | XP_006722694.1 | 4 | 734 | |||
GenBank mRNA | BC002443.1 | - | 4 | 792 | 3346273 | |
BC018831.1 | - | 4 | 792 | |||
U41804.1 | - | 4 | 875 |