Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172654.2 | NP_001166125.1 | 4 | 333 | ||
NM_006865.4 | NP_006856.3 | 4 | 377 | |||
GenBank mRNA | AF014923.1 | - | 4 | 285 | ||
AF014924.1 | - | 4 | 285 | |||
AF025527.1 | - | 4 | 378 | |||
BC028208.1 | - | 4 | 350 | 5215423 | ||
U91926.1 | - | 4 | 347 | |||
U91927.1 | - | 2 | 228 |