Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198595.1 | NP_001185524.1 | 5 | 2567 | ||
NM_006873.3 | NP_006864.2 | 4 | 2450 | |||
GenBank mRNA | AF255310.1 | - | 3 | 2365 | ||
BC121063.2 | - | 3 | 2365 | 40118538 | ||
BC121064.2 | - | 3 | 2365 | 40118539 | ||
CR936807.1 | - | 5 | 2550 |