Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198595.1 | NP_001185524.1 | 5 | 2904 | ||
NM_006873.3 | NP_006864.2 | 4 | 2787 | |||
GenBank mRNA | AL700433.1 | - | 1 | 282 | ||
AL703296.1 | - | 1 | 84 | |||
CR936807.1 | - | 5 | 2887 |