Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027386.2 | 3, 4 | 377 | |||
GenBank mRNA | AK289965.1 | - | 2, 3 | 349 | ||
BT006760.1 | - | 1, 2 | 52 | |||
DT218991.1 | - | 3, 4 | 297 |
Gene Symbol: GUSBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003504.3 | 3, 4 | 399 | |||
GenBank mRNA | BC034017.1 | - | 3, 4 | 400 | ||
BC067351.1 | - | 1, 2 | 97 | |||
BC140728.1 | - | 2, 3 | 222 | |||
BC171739.1 | - | 2, 3 | 222 | 9052954 |
Gene Symbol: GUSBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033968.1 | 1, 2 | 109 | |||
GenBank mRNA | AW500063.1 | - | 1, 2 | 42 | ||
BC104473.1 | - | 1, 2 | 109 | |||
CF141910.1 | - | 1, 2 | 115 |
Gene Symbol: SMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029426.1 | 3, 4 | 295 | |||
GenBank mRNA | AK094021.1 | - | 4, 5 | 701 | ||
AK097091.1 | - | 4, 5 | 675 | |||
AK289832.1 | - | 3, 4 | 377 | |||
AK315849.1 | - | 3, 4 | 368 | |||
BC131740.1 | - | 1, 2 | 109 | |||
X83300.1 | - | 1, 2 | 118 |
Gene Symbol: SMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034021.1 | 4, 5 | 371 | |||
GenBank mRNA | AK289851.1 | - | 3, 4 | 297 | ||
CN312642.1 | - | 1, 2 | 161 | |||
X83301.1 | - | 4, 5 | 371 |