Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012216.3 | NP_036348.2 | 8 | 2045 | ||
NM_052817.2 | NP_438112.2 | 8 | 1955 | |||
XM_005262062.4 | XP_005262119.1 | 8 | 6308 | |||
XM_017029239.1 | XP_016884728.1 | 8 | 6218 | |||
GenBank mRNA | AF196481.1 | - | 8 | 1395 | ||
AY625004.1 | - | 7 | 1322 | |||
BC017707.1 | - | 7 | 1349 | 4446324 | ||
BT006663.1 | - | 7 | 1322 | |||
Y18880.1 | - | 8 | 1669 |