Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281443.1 | NP_001268372.1 | 3 | 134 | ||
NM_007000.3 | NP_008931.1 | 3 | 134 | |||
XM_017026225.1 | XP_016881714.1 | 3 | 523 | |||
GenBank mRNA | AF085807.1 | - | 2 | 102 | ||
BC107097.1 | - | 2 | 107 | 40015698 | ||
BC107098.1 | - | 2 | 107 | 40015700 |