Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286990.1 | NP_001273919.1 | 4 | 406 | ||
NM_007001.2 | NP_008932.2 | 4 | 406 | |||
NR_104627.1 | 4 | 406 | ||||
XM_005251673.1 | XP_005251730.1 | 4 | 381 | |||
XM_005251674.1 | XP_005251731.1 | 4 | 381 | |||
XM_005251675.2 | XP_005251732.1 | 4 | 391 | |||
XM_005251676.1 | XP_005251733.1 | 4 | 381 | |||
XM_006716939.2 | XP_006717002.1 | 4 | 391 | |||
GenBank mRNA | AB106537.1 | - | 4 | 339 | ||
AB122077.1 | - | 4 | 351 | |||
AJ005866.1 | - | 2 | 102 | |||
AK307890.1 | - | 4 | 385 | |||
BC100278.1 | - | 4 | 397 | 6056863 | ||
BC113579.1 | - | 5 | 359 | 8322631 | ||
BX361048.2 | - | 4 | 405 |