Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007011.7 | NP_008942.3 | 10 | 1496 | ||
NM_152924.4 | NP_690888.1 | 6 | 1098 | |||
XM_005254832.1 | XP_005254889.1 | 9 | 1134 | |||
XM_005254833.1 | XP_005254890.1 | 6 | 742 | |||
XM_011521165.1 | XP_011519467.1 | 7 | 947 | |||
GenBank mRNA | AF546700.1 | - | 6 | 830 | ||
AK222841.1 | - | 6 | 770 | |||
AK223083.1 | - | 6 | 768 | |||
AK290340.1 | - | 6 | 770 | |||
BC019248.1 | - | 10 | 1226 | 4826878 | ||
BC090052.1 | - | 6 | 832 | 30388486 | ||
X12433.1 | - | 6 | 753 |