Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007013.3 | NP_008944.1 | 8 | 842 | ||
XM_005250760.3 | XP_005250817.1 | 7 | 803 | |||
XM_005250761.3 | XP_005250818.1 | 8 | 883 | |||
XM_011516802.2 | XP_011515104.1 | 7 | 603 | |||
XM_017012993.1 | XP_016868482.1 | 7 | 795 | |||
XM_017012994.1 | XP_016868483.1 | 7 | 793 | |||
XM_017012995.1 | XP_016868484.1 | 6 | 664 | |||
XM_017012996.1 | XP_016868485.1 | 5 | 582 | |||
XM_017012997.1 | XP_016868486.1 | 5 | 561 | |||
GenBank mRNA | AK292800.1 | - | 8 | 857 | ||
AL136739.1 | - | 6 | 572 | |||
AY043361.1 | - | 7 | 647 | |||
AY345857.1 | - | 8 | 876 | |||
BC036065.1 | - | 8 | 846 | 5296005 |