Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012706.2 | NP_036838.1 | 1 | 234 | ||
GenBank mRNA | X56661.1 | - | 1 | 326 |
Set Membership: |
G-protein coupled receptor |