Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LECT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011705.1 | NP_001011705.1 | 3 | 358 | ||
NM_007015.2 | NP_008946.1 | 3 | 358 | |||
XM_011534897.2 | XP_011533199.1 | 3 | 356 | |||
XM_011534898.2 | XP_011533200.1 | 3 | 357 | |||
XM_011534899.2 | XP_011533201.1 | 3 | 358 | |||
XM_011534900.2 | XP_011533202.1 | 3 | 356 | |||
GenBank mRNA | AY368652.1 | - | 1 | 1175 | ||
BC025659.1 | - | 3 | 358 | 5198878 | ||
CR541910.1 | - | 3 | 247 |