Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013172.1 | NP_037304.1 | 1 | 423 | ||
GenBank mRNA | M27151.1 | - | 1 | 373 |