Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281741.1 | NP_001268670.1 | 5 | 668 | ||
NM_001281742.1 | NP_001268671.1 | 5 | 604 | |||
NM_007019.3 | NP_008950.1 | 6 | 809 | |||
NM_181799.2 | NP_861515.1 | 6 | 755 | |||
NM_181800.2 | NP_861516.1 | 5 | 722 | |||
NM_181801.3 | NP_861517.1 | 6 | 846 | |||
NR_104036.1 | 4 | 517 | ||||
NR_104037.1 | 5 | 612 | ||||
GenBank mRNA | BC007656.2 | - | 6 | 732 | 3345575 | |
BC016292.1 | - | 6 | 738 | 3910286 | ||
BC028161.1 | - | 5 | 1348 | |||
BC050736.1 | - | 6 | 759 | 6503915 | ||
BM556795.1 | - | 6 | 673 | |||
BU169315.1 | - | 6 | 698 | |||
BU171488.1 | - | 5 | 628 | |||
BU844974.1 | - | 5 | 624 | |||
CB143441.1 | - | 4 | 435 | |||
CN354813.1 | - | 5 | 505 | |||
U73379.1 | - | 6 | 729 |