Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034916.2 | NP_001030088.2 | 564 | |||
GenBank mRNA | BC126063.1 | - | 554 | 7307697 | ||
CF111102.1 | - | 564 | ||||
FQ217739.1 | - | 563 | ||||
FQ225104.1 | - | 564 | ||||
FQ226443.1 | - | 562 | ||||
FQ226648.1 | - | 562 | ||||
FQ230224.1 | - | 566 | ||||
FQ230609.1 | - | 571 | ||||
FQ231472.1 | - | 565 | ||||
FQ234329.1 | - | 571 | ||||
X62145.1 | - | 564 |
Gene Symbol: LOC100360117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729143.4 | XP_002729189.1 | 1 | 577 |
Gene Symbol: LOC100910370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750368.3 | XP_003750416.1 | 5 | 643 |