Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017200.1 | NP_058896.1 | 7 | 829 | ||
XM_006234440.2 | XP_006234502.1 | 8 | 1124 | |||
XM_006234441.2 | XP_006234503.1 | 8 | 2145 | |||
XM_006234444.2 | XP_006234506.1 | 8 | 2158 | |||
XM_008761980.1 | XP_008760202.1 | 8 | 1143 | |||
XM_008761981.1 | XP_008760203.1 | 8 | 907 | |||
XM_008761982.1 | XP_008760204.1 | 8 | 1137 | |||
XM_008761983.1 | XP_008760205.1 | 8 | 2160 | |||
XM_008761984.1 | XP_008760206.1 | 8 | 2173 | |||
GenBank mRNA | D10926.1 | - | 7 | 829 | ||
FQ219876.1 | - | 5 | 535 | |||
FQ220339.1 | - | 5 | 529 | |||
FQ220655.1 | - | 5 | 626 | |||
FQ220711.1 | - | 5 | 626 | |||
FQ229364.1 | - | 5 | 558 |