Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317163.1 | NP_001304092.1 | 22 | 3185 | ||
NM_001735.2 | NP_001726.2 | 22 | 2859 | |||
XM_011518980.2 | XP_011517282.1 | 23 | 3228 | |||
XM_017015102.1 | XP_016870591.1 | 24 | 3380 | |||
XM_017015103.1 | XP_016870592.1 | 22 | 3013 | |||
XM_017015104.1 | XP_016870593.1 | 24 | 3380 | |||
GenBank mRNA | AB209031.1 | - | 22 | 3185 | ||
BC113738.1 | - | 22 | 2854 | 8322790 | ||
BC113740.1 | - | 22 | 2854 | 8322792 | ||
M57729.1 | - | 22 | 2841 | |||
M65134.1 | - | 12 | 1596 |