Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KERA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007035.3 | NP_008966.1 | 2 | 1412 | ||
GenBank mRNA | AF063301.1 | - | 1 | 801 | ||
AF205403.1 | - | 2 | 1041 | |||
AK314990.1 | - | 2 | 974 | |||
BC032667.2 | - | 1 | 802 | 5541591 |