Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135604.1 | NP_001129076.1 | 1 | 359 | ||
NM_007036.4 | NP_008967.1 | 1 | 359 | |||
GenBank mRNA | AJ973643.1 | - | 1 | 306 | ||
AK300634.1 | - | 1 | 366 | |||
AK311817.1 | - | 1 | 366 | |||
BC011989.1 | - | 1 | 349 | 3882426 | ||
BM423977.1 | - | 1 | 436 | |||
X89426.1 | - | 1 | 345 |