Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HNRNPUL1 SIRNA Assays ›
Gene Symbol:
HNRNPUL1
Gene Name:
heterogeneous nuclear ribonucleoprotein U like 1
Gene Aliases:
E1B-AP5, E1BAP5, HNRPUL1
Chromosome Location:
Chr.19: 41262476 - 41307783 on Build GRCh38
UniGene ID:
Hs.155218
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s21884
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HNRNPUL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301016.2
NP_001287945.1
10
1501
NM_001321208.1
NP_001308137.1
10
1541
NM_001321211.1
NP_001308140.1
10
1325
NM_007040.5
NP_008971.2
10
1832
NM_144732.4
NP_653333.1
10
1314
XM_005258459.3
XP_005258516.1
10
1635
XM_005258461.4
XP_005258518.1
10
1465
XM_005258463.3
XP_005258520.1
10
1456
XM_005258464.3
XP_005258521.1
10
1271
XM_005258465.4
XP_005258522.1
10
1482
XM_011526392.2
XP_011524694.1
10
1635
XM_011526393.2
XP_011524695.1
10
1557
XM_011526394.2
XP_011524696.1
10
1312
XM_011526395.2
XP_011524697.1
10
1521
GenBank mRNA
AJ007509.1
-
10
1675
AK001229.1
-
1
98
AK022863.1
-
10
1634
AK023006.1
-
10
1271
AK127057.1
-
9
1292
AK225850.1
-
10
1294
AK290729.1
-
10
1296
AK291315.1
-
11
1581
AK291762.1
-
10
1634
AK297127.1
-
10
1429
AK308156.1
-
11
1314
AL050146.1
-
3
440
BC002564.2
-
10
1572
3139049
BC009988.2
-
10
1478
4127873
BC014232.2
-
10
1453
4558792
BC027713.2
-
10
1555
4123077
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
transcription, DNA-templated
regulation of transcription, DNA-templated
RNA processing
response to virus
RNA binding
protein binding
enzyme binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9bfc7:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline