Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007043.6 | NP_008974.5 | 7, 8 | 870 | ||
GenBank mRNA | AK313687.1 | - | 7, 8 | 833 | ||
BC005225.1 | - | 7, 8 | 865 | |||
BC014034.2 | - | 7, 8 | 846 | |||
BC016778.1 | - | 7, 8 | 844 | 4106298 | ||
BC026107.2 | - | 7, 8 | 853 | 4827704 | ||
U55766.1 | - | 7, 8 | 840 |