Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFR1OP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278690.1 | NP_001265619.1 | 5 | 478 | ||
NM_007045.3 | NP_008976.1 | 5 | 478 | |||
NM_194429.2 | NP_919410.1 | 5 | 478 | |||
GenBank mRNA | AJ420472.1 | - | 5 | 374 | ||
AK289846.1 | - | 5 | 401 | |||
AK294950.1 | - | 5 | 401 | |||
AK312791.1 | - | 5 | 389 | |||
BC011902.2 | - | 5 | 391 | 4120239 | ||
DC418208.1 | - | 5 | 479 | |||
Y18046.1 | - | 5 | 467 |