Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZWINT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005413.1 | NP_001005413.1 | 9 | 942 | ||
NM_007057.3 | NP_008988.2 | 9 | 1083 | |||
NM_032997.2 | NP_127490.1 | 8 | 1274 | |||
XM_017015605.1 | XP_016871094.1 | 8 | 1266 | |||
GenBank mRNA | AF067656.1 | - | 9 | 1070 | ||
AK093808.1 | - | 7 | 1384 | |||
AK225509.1 | - | 9 | 1069 | |||
AK293416.1 | - | 8 | 1263 | |||
AK298148.1 | - | 9 | 938 | |||
BC000411.1 | - | 5 | 886 | |||
BC020979.1 | - | 9 | 1070 | 3848873 | ||
BC110399.1 | - | 9 | 1082 | 5554405 |