Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC42EP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152243.2 | NP_689449.1 | 3 | 1527 | ||
XM_006724107.1 | XP_006724170.1 | 3 | 1525 | |||
XM_006724108.1 | XP_006724171.1 | 3 | 1531 | |||
XM_006724109.1 | XP_006724172.1 | 3 | 1491 | |||
XM_017028541.1 | XP_016884030.1 | 3 | 1542 | |||
GenBank mRNA | AK292190.1 | - | 4 | 1472 | ||
BC009356.2 | - | 4 | 1476 | 4300677 | ||
CR456524.1 | - | 2 | 1266 | |||
M88338.1 | - | 3 | 1472 |