Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PWP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317962.1 | NP_001304891.1 | 4 | 388 | ||
NM_001317963.1 | NP_001304892.1 | 4 | 532 | |||
NM_007062.2 | NP_008993.1 | 4 | 532 | |||
GenBank mRNA | AK223162.1 | - | 4 | 456 | ||
AK290681.1 | - | 4 | 373 | |||
AK296252.1 | - | 4 | 389 | |||
BC000067.2 | - | 4 | 415 | |||
BC001652.2 | - | 4 | 451 | 2924215 | ||
BC010921.1 | - | 4 | 466 | |||
BC032127.2 | - | 4 | 454 | |||
BC040135.1 | - | 4 | 430 | |||
BC046144.1 | - | 4 | 446 | |||
DC380488.1 | - | 4 | 532 | |||
L07758.1 | - | 4 | 451 | |||
Z36804.1 | - | 1 | 34 | |||
Z36805.1 | - | 1 | 33 |