Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PWP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317962.1 | NP_001304891.1 | 11 | 1049 | ||
NM_001317963.1 | NP_001304892.1 | 11 | 1221 | |||
NM_007062.2 | NP_008993.1 | 11 | 1193 | |||
GenBank mRNA | AK223162.1 | - | 11 | 1117 | ||
AK290681.1 | - | 11 | 1034 | |||
AK296252.1 | - | 11 | 1050 | |||
AK297961.1 | - | 10 | 878 | |||
BC001652.2 | - | 11 | 1112 | 2924215 | ||
L07758.1 | - | 11 | 1112 |