Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253678.1 | NP_001240607.1 | 1 | 56 | ||
XM_008762543.1 | XP_008760765.1 | 2 | 133 | |||
GenBank mRNA | GQ406386.1 | - | 1 | 56 |